Anti-ODC Rabbit Polyclonal Antibody
ABGEAP14027B
: Abgent
- Antibody type:Primary
- Antigen name:Ornithine Decarboxylase
- Antigen symbol:ODC
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:Ornithine Decarboxylase 1|ODC1|ornithine decarboxylase 1-like|ornithine decarboxylase antizyme inhibitor|AZI
- Size:100 µg
- Pk:400 µl
Type: Primary
Antigen: ODC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human