Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SUMO2 Mouse Monoclonal Antibody (CF405S) [clone: SUMO2/1199]
Anti-SUMO2 Mouse Monoclonal Antibody (CF405S) [clone: SUMO2/1199]
Supplier:  Biotium
Certificates
Anti-SUMO2 Mouse Monoclonal Antibody (CF405S) [clone: SUMO2/1199]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC041199-500
  • BNC041199-100
  • Antigen symbol:
  • Small ubiquitin-related modifier 2
  • Small ubiquitin-related modifier 2
  • Antigen name:
  • SUMO-2
  • SUMO-2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone SUMO2/1199
  • Clone SUMO2/1199
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 474005
  • 474005
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together