Order Entry
United Kingdom
ContactUsLinkComponent
Anti-MCAM Mouse Monoclonal Antibody (CF405S) [clone: MUC18/1130]
Anti-MCAM Mouse Monoclonal Antibody (CF405S) [clone: MUC18/1130]
Supplier:  Biotium
Certificates
Anti-MCAM Mouse Monoclonal Antibody (CF405S) [clone: MUC18/1130]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC041130-100
  • BNC041130-500
  • Antigen symbol:
  • MCAM
  • MCAM
  • Antigen name:
  • MUC18 / Mucin 18 / CD146
  • MUC18 / Mucin 18 / CD146
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone MUC18/1130
  • Clone MUC18/1130
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 599039
  • 599039
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together