Order Entry
United Kingdom
ContactUsLinkComponent
Anti-TOX3 Mouse Monoclonal Antibody (CF568) [clone: TOX3/1123]

Specifications

  • Catalog No:
  • BNC681123-500
  • BNC681123-100
  • Antigen symbol:
  • TOX3
  • TOX3
  • Antigen name:
  • TOX high mobility group box family member 3
  • TOX high mobility group box family member 3
  • Antigen synonyms:
  • CAG trinucleotide repeat-containing gene F9 protein|CAGF9|Trinucleotide repeat containing 9 (TNRC9)|TOX3|TOX high mobility group box family member 3
  • CAG trinucleotide repeat-containing gene F9 protein|CAGF9|Trinucleotide repeat containing 9 (TNRC9)|TOX3|TOX high mobility group box family member 3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone TOX3/1123
  • Clone TOX3/1123
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 460789
  • 460789
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together