Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CDw17 Mouse Monoclonal Antibody [clone: H018.3G-6.F5]
Anti-CDw17 Mouse Monoclonal Antibody [clone: H018.3G-6.F5]
Supplier:  Biotium
Certificates
Anti-CDw17 Mouse Monoclonal Antibody [clone: H018.3G-6.F5]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB1092-500
  • BNUB1092-100
  • Antigen symbol:
  • CDw17
  • CDw17
  • Antigen name:
  • CDw17 antigen
  • CDw17 antigen
  • Antigen synonyms:
  • LacCer|CDw17|)-ceramide|-beta-D-galactosyl-(1-4)--beta-D-glucosyl-(1-1'|Lactosylceramide
  • LacCer|CDw17|)-ceramide|-beta-D-galactosyl-(1-4)--beta-D-glucosyl-(1-1'|Lactosylceramide
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone H018.3G-6.F5
  • Clone H018.3G-6.F5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 162807
  • 162807
  • Isotype:
  • IgM
  • IgM
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together