About this item
Recognizes a protein of 12 kDa, identified as β-microglobulin. β-microglobulin non-covalently associates with the 44 kDa chain to form the HLA Class I antigen complex. Human β-2 microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This MAb is specific to human β-2 microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. The detection of β-2 microglobulin in body fluids has been used as a tumor marker and for monitoring patients with HIV infection.
Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: 246-E9.E7
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC941039-500
- BNC941039-100
- Antigen symbol:
- B2M
- B2M
- Antigen name:
- Beta-2-Microglobulin
- Beta-2-Microglobulin
- Antigen synonyms:
- Hdcma22p|Beta 2 microglobulin|Beta chain of MHC class I molecules|B2M|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin
- Hdcma22p|Beta 2 microglobulin|Beta chain of MHC class I molecules|B2M|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 594
- CF® 594
- Clone:
- Clone 246-E9.E7
- Clone 246-E9.E7
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 534255
- 534255
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide