About this item
At least five CD1 genes (CD1a, b, c, d, and e) are identified. CD1 proteins have been demonstrated to restrict T cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation. CD1a is a non-polymorphic MHC Class 1 related cell surface glycoprotein, expressed in association with Beta-2 microglobulin. Anti-CD1a labels Langerhans cell histiocytosis (Histiocytosis X), extranodal histiocytic sarcoma, a subset of T-lymphoblastic lymphoma/leukemia, and interdigitating dendritic cell sarcoma of the lymph node. When combined with antibodies against TTF-1 and CD5, anti-CD1a is useful in distinguishing between pulmonary and thymic neoplasms since CD1a is consistently expressed in thymic lymphocytes in both typical and atypical thymomas, but only focally in 1/6 of thymic carcinomas and not in lymphocytes in pulmonary neoplasms. Anti-CD1a is reported to be a new marker for perivascular epithelial cell tumor (PEComa).
Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: 66IIC7
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC041034-100
- BNC041034-500
- Antigen symbol:
- CD1A
- CD1A
- Antigen name:
- CD1a molecule
- CD1a molecule
- Antigen synonyms:
- T-Cell Surface Glycoprotein CD1A|FCB6|Cortical thymocyte antigen CD1A|T cell surface antigen T6 / Leu 6|Epidermal dendritic cell marker CD1a|HTA1
- T-Cell Surface Glycoprotein CD1A|FCB6|Cortical thymocyte antigen CD1A|T cell surface antigen T6 / Leu 6|Epidermal dendritic cell marker CD1a|HTA1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- Clone 66IIC7
- Clone 66IIC7
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 1309
- 1309
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide