Order Entry
United Kingdom
ContactUsLinkComponent
Anti-S100A9 Mouse Monoclonal Antibody (Purified) [clone: CPT/1028]
Anti-S100A9 Mouse Monoclonal Antibody (Purified) [clone: CPT/1028]
Supplier:  Biotium
Certificates
Anti-S100A9 Mouse Monoclonal Antibody (Purified) [clone: CPT/1028]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB1028-500
  • BNUB1028-100
  • Antigen symbol:
  • S100A9
  • S100A9
  • Antigen name:
  • Calprotectin
  • Calprotectin
  • Antigen synonyms:
  • calprotectin L1H subunit|MRP14|leukocyte L1 complex heavy chain|migration inhibitory factor-related protein 14|S100-A9|calgranulin-B|S100 calcium-binding protein A9 (calgranulin B)
  • calprotectin L1H subunit|MRP14|leukocyte L1 complex heavy chain|migration inhibitory factor-related protein 14|S100-A9|calgranulin-B|S100 calcium-binding protein A9 (calgranulin B)
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone CPT/1028
  • Clone CPT/1028
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 501497 , 715224
  • 501497 , 715224
  • Isotype:
  • IgM kappa
  • IgM kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together