Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CPS1 Mouse Monoclonal Antibody (CF594) [clone: CPS1/1022]
Anti-CPS1 Mouse Monoclonal Antibody (CF594) [clone: CPS1/1022]
Supplier:  Biotium
Certificates
Anti-CPS1 Mouse Monoclonal Antibody (CF594) [clone: CPS1/1022]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC941022-100
  • BNC941022-500
  • Antigen symbol:
  • CPS1
  • CPS1
  • Antigen name:
  • Hepatocyte Specific
  • Hepatocyte Specific
  • Antigen synonyms:
  • CPSase 1|Carbamoylphosphate synthetase 1|CPSASE1|Carbamoyl-phosphate synthetase 1 (CPS1)
  • CPSase 1|Carbamoylphosphate synthetase 1|CPSASE1|Carbamoyl-phosphate synthetase 1 (CPS1)
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Clone:
  • Clone CPS1/1022
  • Clone CPS1/1022
  • Reactivity:
  • Human, Dog
  • Human, Dog
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 149252
  • 149252
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together