Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SOX10 Mouse Monoclonal Antibody (CF568) [clone: SOX10/1074]
Anti-SOX10 Mouse Monoclonal Antibody (CF568) [clone: SOX10/1074]
Supplier:  Biotium
Certificates
Anti-SOX10 Mouse Monoclonal Antibody (CF568) [clone: SOX10/1074]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC681017-100
  • BNC681017-500
  • Antigen symbol:
  • SOX10
  • SOX10
  • Antigen synonyms:
  • Waardenburg syndrome type 2E (WS2E)|PCWH|WS4|SRY (sex determining region Y) box 10|Waardenburg syndrome type 4C (WS4C)|DOM|SRY box containing gene 10|SOX10|SRY related HMG box gene 10
  • Waardenburg syndrome type 2E (WS2E)|PCWH|WS4|SRY (sex determining region Y) box 10|Waardenburg syndrome type 4C (WS4C)|DOM|SRY box containing gene 10|SOX10|SRY related HMG box gene 10
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone SOX10/1074
  • Clone SOX10/1074
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 376984
  • 376984
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together