Anti-HIF-PH3 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNB100-139B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:HIF Prolyl Hydroxylase 3
- Antigen symbol:HIF-PH3
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:112399
- Antigen synonyms:egl nine homolog 3|prolyl hydroxylase domain-containing protein 3|HIF-PH3|egl nine-like protein 3 isoform|HIF-prolyl hydroxylase 3|HIF prolyl hydroxylase 3|PHD3|HPH-1|HIFPH3|HPH-3|hypoxia-inducible factor prolyl hydroxylase 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Rabbit Polyclonal HIF Prolyl Hydroxylase 3 Antibody [Biotin]. Tested Applications: Western Blot. Tested Reactivity: Human, Rat, Mouse (-).
Type: Primary
Antigen: HIF-PH3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human