Anti-KRT7 Mouse Monoclonal Antibody (CF640R) [clone: KRT7/760 KRT7/903]
: Biotium
This antibody recognizes an intermediate filament protein (IFP) of 55 kDa, which is identified as cytokeratin 7. This MAb is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This MAb is highly useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).
Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: KRT7/760 KRT7/903
Conjugation: CF640R
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgGs
- Catalog No:
- BNC400906-100
- BNC400906-500
- Antigen symbol:
- KRT7
- KRT7
- Antigen name:
- Cytokeratin 7
- Cytokeratin 7
- Antigen synonyms:
- KRT7|Krt2-7|Sarcolectin|SCL|K2C7|Keratin 55K Type II Cytoskeletal|CK-7|Type-II Keratin Kb7|Type II Mesothelial Keratin K7|Keratin Simple Epithelial Type 1 K7|Keratin Type II Cytoskeletal 7
- KRT7|Krt2-7|Sarcolectin|SCL|K2C7|Keratin 55K Type II Cytoskeletal|CK-7|Type-II Keratin Kb7|Type II Mesothelial Keratin K7|Keratin Simple Epithelial Type 1 K7|Keratin Type II Cytoskeletal 7
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone KRT7/760 KRT7/903
- Clone KRT7/760 KRT7/903
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 411501
- 411501
- Isotype:
- IgGs
- IgGs
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Frequently Bought Together