Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-CD1A Mouse Monoclonal Antibody (CF647) [clone: C1A/711]
 :  Biotium
 
Anti-CD1A Mouse Monoclonal Antibody (CF647) [clone: C1A/711]
 :  Biotium

 

  • Catalog No:
  • BNC470711-100
  • BNC470711-500
  • Antigen symbol:
  • CD1A
  • CD1A
  • Antigen name:
  • CD1a molecule
  • CD1a molecule
  • Antigen synonyms:
  • T-Cell Surface Glycoprotein CD1A|FCB6|Epidermal dendritic cell marker CD1a antibody|Cortical thymocyte antigen CD1A|T cell surface antigen T6 / Leu 6|HTA1
  • T-Cell Surface Glycoprotein CD1A|FCB6|Epidermal dendritic cell marker CD1a antibody|Cortical thymocyte antigen CD1A|T cell surface antigen T6 / Leu 6|HTA1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone C1A/711
  • Clone C1A/711
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 1309
  • 1309
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together