Anti-MYC Mouse Monoclonal Antibody (CF555) [clone: 9E10.]
Supplier: Biotium
Certificates
About this item
This antibody recognizes a transcription factor of 64-67 kDa, identified as c-myc. Its epitope spans between aa 410-419 (EQKLISEEDL) which is a specific portion of an alpha helical region of human c-myc protein. This MAb shows no cross-reaction with v-myc. c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors. Over-expression of c-myc protein occurs frequently in luminal cells of prostate intraepithelial neoplasia as well as in most primary carcinomas and metastatic disease.
Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: 9E10.
Conjugation: CF555
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC550596-500
- BNC550596-100
- Antigen symbol:
- MYC
- MYC
- Antigen name:
- c-Myc
- c-Myc
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 555
- CF® 555
- Clone:
- Clone 9E10.
- Clone 9E10.
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 202453
- 202453
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together