Order Entry
United Kingdom
ContactUsLinkComponent
Anti-MYC Mouse Monoclonal Antibody (CF405S) [clone: 9E10.]
Anti-MYC Mouse Monoclonal Antibody (CF405S) [clone: 9E10.]
Supplier:  Biotium
Certificates
Anti-MYC Mouse Monoclonal Antibody (CF405S) [clone: 9E10.]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC040596-500
  • BNC040596-100
  • Antigen symbol:
  • MYC
  • MYC
  • Antigen name:
  • c-Myc
  • c-Myc
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone 9E10.
  • Clone 9E10.
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 202453
  • 202453
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together