Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ODC1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ODC1/486]
Anti-ODC1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ODC1/486]
  BNUM0486-50
 :  Biotium
Anti-ODC1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ODC1/486]
  BNUM0486-50
 :  Biotium

 

  • Antibody type:
    Primary
  • Antigen name:
    ODC-1
  • Antigen symbol:
    ODC1
  • Clonality:
    Monoclonal
  • Clone:
    Clone ODC1/486
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Gene ID:
    467701
  • Antigen synonyms:
    RNODC|Ornithine decarboxylase structural 1
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

 

 

Recognizes a 53 kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.

Type: Primary
Antigen: ODC1
Clonality: Monoclonal
Clone: ODC1/486
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Rat