Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ODC1 Mouse Monoclonal Antibody (CF488A) [clone: ODC1/486]
Anti-ODC1 Mouse Monoclonal Antibody (CF488A) [clone: ODC1/486]
Supplier:  Biotium
Certificates
Anti-ODC1 Mouse Monoclonal Antibody (CF488A) [clone: ODC1/486]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC880486-500
  • BNC880486-100
  • Antigen symbol:
  • ODC1
  • ODC1
  • Antigen name:
  • ODC-1
  • ODC-1
  • Antigen synonyms:
  • RNODC|Ornithine decarboxylase structural 1
  • RNODC|Ornithine decarboxylase structural 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone ODC1/486
  • Clone ODC1/486
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 467701
  • 467701
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together