CD19 is a transmembrane glycoprotein that contains two extracellular immunoglobulin-like domains. CD19 is present in both benign and malignant B-cells and is considered to be the most reliable surface marker of this lineage over a wide range of maturational stages. In normal lymphoid tissue, CD19 is observed in germinal centers, in mantle zone cells, and in scattered cells of the inter-follicular areas. Anti-CD19 exhibits an overall immunoreactivity pattern similar to those of the antibodies against CD20 and CD22. However, in contrast to CD20, expression of CD19 is continuous throughout B-cell development and through terminal differentiation of B-cells into plasma cells. Anti-CD19 positivity is seen in the vast majority of B-cell neoplasms commonly at a lower intensity than normal B-cell counterparts. Plasma cell neoplasms are nearly always negative, as are T-cell neoplasms.
Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: CVID3/429
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Human, Monkey
- Catalog No:
- BNC880429-100
- BNC880429-500
- Antigen symbol:
- CD19
- CD19
- Antigen synonyms:
- CVID3|Lymphocyte Surface Antigen|Leu-12|B-lymphocyte antigen CD19|T-cell surface antigen Leu-12|B-lymphocyte surface antigen B4
- CVID3|Lymphocyte Surface Antigen|Leu-12|B-lymphocyte antigen CD19|T-cell surface antigen Leu-12|B-lymphocyte surface antigen B4
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone CVID3/429
- Clone CVID3/429
- Reactivity:
- Monkey, Chimpanzee, Human
- Monkey, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 652262
- 652262
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide