Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Biotin Mouse Monoclonal Antibody (CF555) [clone: Hyb8]
Anti-Biotin Mouse Monoclonal Antibody (CF555) [clone: Hyb8]
Supplier:  Biotium
Certificates
Anti-Biotin Mouse Monoclonal Antibody (CF555) [clone: Hyb8]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC550400-100
  • BNC550400-500
  • Antigen name:
  • Biotin
  • Biotin
  • Antigen synonyms:
  • Vitamin B7 or Vitamin H
  • Vitamin B7 or Vitamin H
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 555
  • CF® 555
  • Clone:
  • Clone Hyb8
  • Clone Hyb8
  • Reactivity:
  • Multiple
  • Multiple
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together