Order Entry
United Kingdom
ContactUsLinkComponent
Anti-VCAM1 Mouse Monoclonal Antibody (CF647) [clone: 1.4C3]
Anti-VCAM1 Mouse Monoclonal Antibody (CF647) [clone: 1.4C3]
Supplier:  Biotium
Certificates
Anti-VCAM1 Mouse Monoclonal Antibody (CF647) [clone: 1.4C3]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470149-100
  • BNC470149-500
  • Antigen symbol:
  • VCAM1
  • VCAM1
  • Antigen name:
  • Vascular cell adhesion molecule 1
  • Vascular cell adhesion molecule 1
  • Antigen synonyms:
  • CD106|Vascular cell adhesion protein 1|INCAM-100|Vascular Cell Adhesion Molecule 1 (VCAM1)|L1CAM
  • CD106|Vascular cell adhesion protein 1|INCAM-100|Vascular Cell Adhesion Molecule 1 (VCAM1)|L1CAM
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone 1.4C3
  • Clone 1.4C3
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 109225
  • 109225
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together