Anti-CRYAB Mouse Monoclonal Antibody [clone: 1B6.1-3G4]
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Alpha-crystallins, which are part of the small Heat shock family members, are major water-soluble proteins present in the lens of the mammalian eye.
- Dilution range: Immunohistochemistry (paraffin sections, 10 µg/ml), Western blot (1:1,000, colorimetric)
- Detects a band of ~20kDa by Western blot
Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 1B6.1-3G4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Chicken, Human, Mouse, Rat
Specifications
- Catalog No:
- ENZOADISPA222F
- ENZOADISPA222D
- Antigen symbol:
- CRYAB
- CRYAB
- Antigen name:
- Crystallin Alpha B
- Crystallin Alpha B
- Antigen synonyms:
- Heat-shock protein beta-5|HSPB5|CRYA2|Alpha Crystallin B chain|Alpha (B)-Crystallin|CryAB|Rosenthal fiber componet|CTPP2
- Heat-shock protein beta-5|HSPB5|CRYA2|Alpha Crystallin B chain|Alpha (B)-Crystallin|CryAB|Rosenthal fiber componet|CTPP2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1B6.1-3G4
- 1B6.1-3G4
- Reactivity:
- Chicken, Human, Bovine, Rat, Mouse
- Chicken, Human, Bovine, Rat, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Bovine alphaB-crystallin
- Bovine alphaB-crystallin
- Form:
- Liquid
- Liquid
- Purification:
- Purified by Protein G affinity chromatography
- Purified by Protein G affinity chromatography
- Storage buffer:
- PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
- PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together