Anti-TXN Mouse Monoclonal Antibody [clone: 2C9]
Catalog # ENZOADIMSA150E
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Thioredoxin 1
- Antigen symbol:TXN1
- Clonality:Monoclonal
- Clone:2C9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:TRDX|TRX|TRX1
- Storage buffer:PBS containing 50% glycerol
- Storage temperature:−20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:E. coli thioredoxin (trxA)
- Purification:Purified by Protein A affinity chromatography
- Pk:100 µG
Specifications
About this item
Thioredoxins act as antioxidants by reducing the proteins by cysteine thiol-disulfide exchange. Several plasmid expression vectors have been constructed that direct the synthesis of foreign polypeptides in E. coli as fusions with C-terminal Thioredoxin (TrxA) to offer soluble expression of normally insoluble or difficult to express proteins such as cytokines and growth factors.
- Western blot (1 µg/ml, ECL)
- Recognizes trx and trx-tag-f thioredoxin
Type: Primary
Antigen: TXN
Clonality: Monoclonal
Clone: 2C9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: E. coli