Zinc chloride, white crystalline powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7646-85-7
:
Formula:ZnCl₂
MDL Number:MFCD00011295
:136.3 g/mol
Melting Pt:290 °C (lit.)
UN:2331
ADR:8,III
:GHS05
:GHS07
:GHS09
Specification Test Results
- Assay:>97,0%
- Oxychloride:Passes Test
- Insoluble Matter:≤0,005%
- Nitrate (NO3):≤0,003%
- Sulfate (SO4):≤0,01%
- Ammonium (NH4):≤0,005%
- Calcium (Ca):≤0,06%
- Iron (Fe):≤0,001%
- Lead (Pb):≤0,005%
- Magnesium (Mg):<0,01%
- Potassium (K):<0,02%
- Sodium (Na):<0,05%
Specification Test Results
Frequently Bought Together