Order Entry
United Kingdom
ContactUsLinkComponent
Dyclonine hydrochloride, white powder, fast flow
Dyclonine hydrochloride, white powder, fast flow
Catalog # ICNA0219019425
Supplier:  MP Biomedicals
CAS Number:  
Dyclonine hydrochloride, white powder, fast flow
Catalog # ICNA0219019425
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Dyclonine HCl, 1-(4-Butoxyphenyl)-3-(piperidin-1-yl)propan-1-one hydrochloride
 :
 :325.877 g/mol
CAS Number:536-43-6
Formula:C₁₈H₂₈ClNO₂
MDL Number:MFCD00035386