
Indocyanine green ~90% (dye content)
:3599-32-4
:
:C₄₃H₄₇N₂NaO₆S₂
:MFCD00013078
:774.99 g/mol
:222-751-5
:GHS07
:Ambient
- Identity Test:Passes
Frequently Bought Together
2-Propanol, EMPROVE® EXPERT Ph. Eur., BP, USP for preparative chromatography, SAFC®
: MERCK PRODUCTION CHEMICALS
KOLLIPHOR RH 40 1 * 1 kg
: Merck
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%
: Thermo Fisher Scientific
Thermo-tube (gamma-irradiated) natural (AB-0532) 1 * 1.000 items
: Thermo Fisher Scientific
Butanone (Methylethylketone) pure 1 * 2,5 L
: PanReac AppliChem
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Sodium hexanitritocobaltate(III)
: Thermo Fisher Scientific
RS pro storage bin, 130 mm x 205 mm blue, 5 pack 1 * 5 items
: RS COMPONENTS
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
: Thermo Fisher Scientific
Microcentrifuge tubes, 0,6/1,5/2,0 ml, graduated, with snap cap
: Thermo Fisher Scientific
Colour coders for cryovials, CryoColorCode, Nunc™
: Thermo Fisher Scientific
Chloroform 99+% stabilised, extra pure
: Thermo Fisher Scientific