Cinnarizine, white powder
Catalog # ICNA0215498625
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:368.524 g/mol
Storage Temperature:Ambient
CAS Number:298-57-7
Formula:C₂₆H₂₈N₂
MDL Number:MFCD00056037