Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Oligomycin

Oligomycin

Supplier:   MP Biomedicals
Oligomycin
Supplier:  MP Biomedicals
Certificates

3 Options Available Below

Product Details & Documents

Soluble in ether, benzene, absolute ethanol, glacial acetic acid and acetone.

Synonyms:  Oligomycin A, B, C, Oligomycin complex
  • CAS Number:1404-19-9
  • Formula:C₄₅H₇₄O₁₁
  • MDL Number:MFCD01779388
  • Molecular Weight:789.3 g/mol
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - £45.90
    £45.90
    1 * 1 mg
       £45.90

  • 5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - £128.00
    £128.00
    1 * 5 mg
       £128.00

  • 10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - £233.00
    £233.00
    1 * 10 mg
       £233.00