
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]), light green powder
Catalog # ICNA0219502305
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- UV/VIS: I max (water):340 - 342, 224 - 228 nm
- Moisture:≤5%
Specification Test Results
About this item
:
:548.68 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:30931-67-0
Formula:C₁₈H₂₄N₆O₆S₄
MDL Number:MFCD00010404