1-Pentanesulphonic acid sodium salt monohydrate, fine crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Sodium 1-pentanesulphonate monohydrate
CAS Number:207605-40-1
:MFCD00149548
Formula:C₅H₁₁NaO₃S·1H₂O
MDL Number:MFCD00007541
Molecular Weight:192.21 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Moisture:7 - 9%
Specification Test Results
Frequently Bought Together