trans-4-Hydroxycinnamic acid ≥98%, powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:501-98-4
:7400-08-0
Formula:C₉H₈O₃
MDL Number:MFCD00004399
:164.16 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥98%
- FTIR:Conforms to Standard
- Melting Point:212 - 222 °C
Specification Test Results
Frequently Bought Together