Anti-SIGLEC7 Mouse Polyclonal Antibody
Catalog # ABNOH00027036-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 7
- Antigen symbol:SIGLEC7
- Clonality:Polyclonal
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Epitope:Full-length
- Gene ID:27036
- Sequence:MLLLLLLPLLWGRERVEGQKSNRKDYSLTMQSSVTVQEGMCVHVRCSFSYPVDSQTDSDPVHGYWFRAGNDISWKAPVATNNPAWAVQEETRDRFHLLGDPQTKNCTLSIRDARMSDAGRYFFRMEKGNIKWNYKYDQLSVNVTALTHRPNILIPGTLESGCFQNLTCSVPWACEQGTPPMISWMGTSVSPLHPSTTRSSVLTLIPQPQHHGTSLTCQVTLPGAGVTTNRTIQLNVSYPPQNLTVTVFQGEGTASTALGNSSSLSVLEGQSLRLVCAVDSNPPARLSWTWRSLTLYPSQPSNPLVLELQVHLGDEGEFTCRAQNSLGSQHVSLNLSLQQEYTGKMRPVSGVLLGAVGGAGATALVFLSFCVIFIVVRSCRKKSARPAADVGDIGMKDANTIRGSASQGNLTESWADDNPRHHGLAAHSSGEEREIQYAPLSFHKGEPQDLSGQEATNNEYSEIKIPK
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human SIGLEC7 protein.
Type: Primary
Antigen: SIGLEC7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Full-length
Host: Mouse
Isotype:
Reactivity: Human