Anti-BrdU Mouse Monoclonal Antibody [clone: BRD.3]
Catalog # ABNOMAB13237
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Bromodeoxyuridine (BrdU)
- Antigen symbol:BrdU
- Clonality:Monoclonal
- Clone:BRD.3
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against Bromodeoxyuridine (BrdU).
Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: BRD.3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: