Anti-VERO Rabbit Polyclonal Antibody (FITC) [clone: ]
Catalog # ABNOPAB29824
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:VERO cells
- Antigen symbol:VERO
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Isotype:IgG
- Reactivity:African green monkey
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody raised against VERO cells.
Type: Primary
Antigen: VERO
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: African green monkey