Order Entry
United Kingdom
ContactUsLinkComponent
 
Ultra-low temperature chest freezers, −85 °C
 
undefined
Ultra-low temperature chest freezers, −85 °C

 

  • Catalog No:
  • DAIRULTF320
  • DAIRULTF80
  • DAIRULTF420
  • Model:
  • ULTF 320
  • ULTF 80
  • ULTF 420
  • Cat. No.:
  • DAIRULTF320
  • DAIRULTF80
  • DAIRULTF420
  • Display:
  • Digital
  • Digital
  • Digital
  • Alarm:
  • Audible and visual
  • Audible and visual
  • Audible and visual
  • Noise level:
  • <50 dB (A)
  • <50 dB (A)
  • <50 dB (A)
  • Access ports:
  • 1
  • 1
  • 1
  • Weight:
  • 89 kg
  • 45 kg
  • 100 kg

 

Frequently Bought Together