Fluorescent cell counters, Cellometer® X
Supplier: NEXCELOM BIOSCIENCE
About this item
The Cellometer X are PC-based cell imagers for fluorescent image capture and analysis.
- Cell count and viability in less than 30 seconds
- Ability to perform cell-based assays
- Cell size range 0,5 to 15 µm and sample volume 20 µl
Both can perform cell counting, viability, intensity and population analysis, for yeast concentration in brewing fermentations.
: Supplied with the Cellometer® software.
2 Options Available Below
Product Details & Documents
The Cellometer X are PC-based cell imagers for fluorescent image capture and analysis.
- Cell count and viability in less than 30 seconds
- Ability to perform cell-based assays
- Cell size range 0,5 to 15 µm and sample volume 20 µl
Both can perform cell counting, viability, intensity and population analysis, for yeast concentration in brewing fermentations.
: Supplied with the Cellometer® software.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- NEXCMX2
- NEXCCMX2-SK150
- Cell size:
- 0,5 − 15 µm
- 0,5 − 15 µm
- Sample volume:
- 20 µl
- 20 µl
- Measurement time:
- 30 s
Specifications
Product Family Options
Product Information
- ModelAvailabilityPrice
- NEXCMX2Fluorescent cell counter with one brightfield and dual fluorescent channels (red and green)Fluorescent cell counter with one brightfield and dual fluorescent channels (red and green)X2Log in for availability1 * 1 items£19,200.00Specifications:
More
Specifications
Cat. No.NEXCMX2Cell size0,5 − 15 µmSample volume20 µlMeasurement time30 s - NEXCCMX2-SK150Fluorescent cell counter with one brightfield and dual fluorescent channels (red and green) and PC laptopFluorescent cell counter with one brightfield and dual fluorescent channels (red and green) and PC laptopX2Log in for availability1 * 1 itemsSpecifications:
More
Specifications
Cat. No.NEXCCMX2-SK150Cell size0,5 − 15 µmSample volume20 µlMeasurement time
Accessories
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambersDisposable plastic counting chambersNEXCCHT4-PD100-002Log in for availability1 * 100 items - £523.00
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambers
Product Information
- Catalog #AvailabilityPrice
- Laptop computer for Cellometer®
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambers
Product Information
- Catalog #AvailabilityPrice
- Disposable plastic counting chambers
Product Information
- Catalog #AvailabilityPrice
- Validation solution for Cellometer® fluorescent instruments (100 tests) in a vialValidation solution for Cellometer® fluorescent instruments (100 tests) in a vialNEXCCCBM-011-2MLLog in for availability1 * 2 mL - £619.00



