Order Entry
United Kingdom
ContactUsLinkComponent
5-Norbornene-2-carboxylic acid 98% (mixture of isomers)
Corrosive
5-Norbornene-2-carboxylic acid 98% (mixture of isomers)
Certificates
undefined
Corrosive
5-Norbornene-2-carboxylic acid 98% (mixture of isomers)

Specification Test Results

  • Appearance:
    Colorless to light yellow Liquid or solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4925 to 1.4965 (20°C, 589 nm)
  • GC:
    ≥97.5 % (total of isomers)
  • Titration with NaOH:
    97.5 to 102.5 %

Specification Test Results

Related Information

Frequently Bought Together