Potassium hydrogen carbonate 99.5% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:298-14-6
:
Formula:KHCO₃
MDL Number:MFCD00011402
:100.12 g/mol
Melting Pt:292 °C
Density:2,17 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Solubility:Clear colorless
- UV:(1 M in water)
- Titration with HClO4:99.4 to 100.6 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO4):≤0.005 %
- Calcium (Ca):≤0.001 %
- Iron (Fe):≤0.0005 %
- Sodium (Na):≤0.02 %
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together