Order Entry
United Kingdom
ContactUsLinkComponent
 
Irritant
3-Amino-4-fluorophenylboronic acid 97%
Catalog # ACRO445290010
CAS Number:  
undefined
Irritant
3-Amino-4-fluorophenylboronic acid 97%
Catalog # ACRO445290010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to light brown Powder or solid
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

 :
 :154.94 g/mol
Melting Pt:226…232 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:873566-75-7
Formula:C₆H₇BFNO₂
MDL Number:MFCD03092925