Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
N'-Hydroxy-2,2-dimethylpropanimidamide 95%

N'-Hydroxy-2,2-dimethylpropanimidamide 95%

Irritant
N'-Hydroxy-2,2-dimethylpropanimidamide 95%
Certificates
N'-Hydroxy-2,2-dimethylpropanimidamide 95%

2 Options Available Below

Product Details & Documents
Synonyms:  N-Hydroxypivalimidamide, (1Z)-N'-hydroxy-2,2-dimethylpropanimidamide, Propanimidamide, N'-hydroxy-2,2-dimethyl-
  • CAS Number:42956-75-2
  • Formula:C₅H₁₂N₂O
  • MDL Number:MFCD03426250
  • Molecular Weight:116.16 g/mol
  • Melting Pt:108.6…113.7 °C
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥94.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - £65.60
    £65.60
    1 * 1 g
       £65.60

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £197.00
    £197.00
    1 * 5 g
       £197.00