
2-Iodo-5-methylthiophene 97%
ACRO439270010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:Yellow to orange-red Liquid
- Infrared spectrum:Conforms
- GC:≥96.5 %
:
:224.07 g/mol
:81…83 °C (10 mmHg)
:25…28 °C
:1.852 g/cm³
:89 °C
:GHS07
:Ambient
:16494-36-3
:C₅H₅IS
:MFCD00022492