Order Entry
United Kingdom
ContactUsLinkComponent
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt) 95%
Irritant
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt) 95%
Certificates
undefined
Irritant
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt) 95%

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Sodium (Na):
    9.95 to 11.22 %
  • Elemental Analysis:
    ≥6.06 to ≤6.84 % Nitrogen

Specification Test Results

Related Information

Frequently Bought Together