



Methylzinc chloride 2 M in THF, AcroSeal®
Catalog # 436080500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Turbid colorless to light yellow Solution
- Titration Argentometric:1.90 to 2.20 M
- GC:≥97.5 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:115.865 g/mol
Density:1.033 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:5158-46-3
Formula:CH₃ClZn
MDL Number:MFCD00192334