
6-Amino-3-methyl-3H-benzooxazol-2-one 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99584-10-8
:
Formula:C₈H₈N₂O₂
MDL Number:MFCD07801206
:164.17 g/mol
Melting Pt:148…151 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Tan Powder
- Infrared spectrum:Conforms
- Melting point:148 to 152 °C
- HPLC:≥88.0 %
Specification Test Results
Frequently Bought Together