
3-Bromo-5-(trifluoromethyl)pyridin-2-ol 97%
Catalog # ACRO432560050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:168 to 172 °C
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:241.995 g/mol
Melting Pt:170.1…171.2 °C
:GHS07
Storage Temperature:Ambient
CAS Number:76041-73-1
Formula:C₆H₃BrF₃NO
MDL Number:MFCD02691223