Benzo[c][1,2,5]thiadiazole-5-boronic acid pinacol ester 97%
Catalog # ACRO430570010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Brown Powder
- Infrared spectrum:Conforms
- Melting point:83 to 87 °C
- GC:≥96.0 %
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:262.134 g/mol
Melting Pt:84…86 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1168135-03-2
Formula:C₁₂H₁₅BN₂O₂S
MDL Number:MFCD11867874