Order Entry
United Kingdom
ContactUsLinkComponent
Amberlite® IRN-78, Ion exchange resin cationic type (OH-) ion-exchange resin
Amberlite® IRN-78, Ion exchange resin cationic type (OH-) ion-exchange resin
Certificates
undefined
Amberlite® IRN-78, Ion exchange resin cationic type (OH-) ion-exchange resin
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Yellow to amber Spherical beads
  • Harmonic mean size:
    0.580 to 0.680 mm
  • Uniformity coefficient:
    ≤1.20
  • Moisture holding capacity:
    54.0 to 60.0 % (OH)
  • Total volume capacity:
    ≥1.10 eq/l (OH) as is