cis-4,7,10,13,16,19-Docosahexaenoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: DHA, Docosa-4Z,7Z,10Z,13Z,16Z,19Z-Hexaenoic Acid
CAS Number:6217-54-5
:
Formula:C₂₂H₃₂O₂
MDL Number:MFCD00065722
Density:0.95 g/cm³
Molecular Weight:328.50 g/mol
Melting Pt:–44 °C
Flash Pt:62 °C
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together