1-Iodo-3,3,3-trifluoropropane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,1,1-Trifluoro-3-iodopropane, 3-Iodo-1,1,1-trifluoropropane
CAS Number:460-37-7
:
Formula:C₃H₄F₃I
MDL Number:MFCD00038531
Boiling Pt:89…91 °C (760 mmHg)
Density:1.911 g/cm³
Molecular Weight:223.96 g/mol
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to pink Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4190 to 1.4210 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together