Sodium methanesulphinate 85%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Methanesulphinic acid, sodium salt
- CAS Number:20277-69-4
- Formula:CH₃NaO₂S
- MDL Number:MFCD00040392
- Molecular Weight:102.09 g/mol
- Melting Pt:222…226 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: Methanesulphinic acid, sodium salt
- CAS Number:20277-69-4
- Formula:CH₃NaO₂S
- MDL Number:MFCD00040392
- Molecular Weight:102.09 g/mol
- Melting Pt:222…226 °C
- Storage Temperature:Ambient
Documents
ACRO39296
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥80 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



