
4-(Bromomethyl)phenylboronic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:68162-47-0
:
Formula:C₇H₈BBrO₂
MDL Number:MFCD01318113
Melting Pt:165…172 °C (dec.)
ADR:8,III
:214.85
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together